Assay Details
| Gene Symbol: march5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123478.1 | NP_001116950.1 | 3 - 4 | 541 | 66 | |
| GenBank mRNA | BC160533.1 | - | 3 - 4 | 541 | 66 | |
More Information
Set Membership: |
|

march5
Genomic Map