Assay Details
| Gene Symbol: rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126497.1 | NP_001119969.1 | 4 - 5 | 672 | 68 | |
| XM_012970048.2 | XP_012825502.1 | 4 - 5 | 757 | 68 | ||
| XM_012970049.2 | XP_012825503.1 | 4 - 5 | 764 | 68 | ||
| XM_018093780.1 | XP_017949269.1 | 4 - 5 | 640 | 68 | ||
| GenBank mRNA | BC158160.1 | - | 4 - 5 | 672 | 68 | |
More Information
Set Membership: |
|

rnf168
Genomic Map