Assay Details
| Gene Symbol: syp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126502.1 | NP_001119974.1 | 2 - 3 | 201 | 75 | |
| GenBank mRNA | BC158217.1 | - | 2 - 3 | 201 | 75 | |
| CT030505.1 | - | 133 | 75 | |||
More Information
Set Membership: |
|

syp
Genomic Map