Assay Details
| Gene Symbol: dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126626.1 | NP_001120098.1 | 3 - 4 | 763 | 61 | |
| GenBank mRNA | BC158896.1 | - | 3 - 4 | 763 | 61 | |
| CU025139.1 | - | 3 - 4 | 596 | 61 | ||
More Information
Set Membership: |
|

dohh
Genomic Map