Assay Details
| Gene Symbol: lrwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126658.1 | NP_001120130.1 | 13 | 1901 | 87 | |
| GenBank mRNA | BC159054.1 | - | 13 | 1901 | 87 | |
| CT030321.1 | - | 13 | 2001 | 87 | ||
More Information
Set Membership: |
|

lrwd1
Genomic Map