Assay Details
| Gene Symbol: fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126664.1 | NP_001120136.1 | 1 - 2 | 100 | 101 | |
| GenBank mRNA | BC159080.1 | - | 1 - 2 | 100 | 101 | |
| CT025469.2 | - | 1 - 2 | 96 | 101 | ||
More Information
Set Membership: |
|

fam32a
Genomic Map