Assay Details
| Gene Symbol: nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126669.1 | NP_001120141.1 | 6 - 7 | 1696 | 67 | |
| XM_012956444.2 | XP_012811898.2 | 6 - 7 | 1358 | 67 | ||
| XM_012956446.2 | XP_012811900.1 | 6 - 7 | 1781 | 67 | ||
| GenBank mRNA | BC159129.1 | - | 6 - 7 | 1696 | 67 | |
More Information
Set Membership: |
|

nab2
Genomic Map