Assay Details
| Gene Symbol: lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126727.1 | NP_001120199.1 | 4 - 5 | 642 | 68 | |
| XM_012968002.2 | XP_012823456.1 | 4 - 5 | 724 | 68 | ||
| XM_012968003.2 | XP_012823457.1 | 4 - 5 | 1067 | 68 | ||
| GenBank mRNA | BC159320.1 | - | 4 - 5 | 642 | 68 | |
More Information
Set Membership: |
|

lrrc57
Genomic Map