Assay Details
| Gene Symbol: smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126885.1 | NP_001120357.1 | 8 - 9 | 1354 | 66 | |
| XM_012956146.2 | XP_012811600.1 | 9 - 10 | 1407 | 66 | ||
| GenBank mRNA | BC161001.1 | - | 8 - 9 | 1354 | 66 | |
More Information
Set Membership: |
|

smyd1
Genomic Map