Assay Details
| Gene Symbol: ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126957.1 | NP_001120429.1 | 10 - 11 | 1132 | 74 | |
| XM_012966087.2 | XP_012821541.1 | 10 - 11 | 1210 | 74 | ||
| XM_012966088.2 | XP_012821542.1 | 10 - 11 | 1211 | 74 | ||
| XM_012966089.2 | XP_012821543.1 | 10 - 11 | 1207 | 74 | ||
| GenBank mRNA | BC161184.1 | - | 10 - 11 | 1132 | 74 | |
More Information
Set Membership: |
|

ccdc63
Genomic Map