Assay Details
| Gene Symbol: ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127005.1 | NP_001120477.1 | 2 - 3 | 206 | 78 | |
| XM_012956495.2 | XP_012811949.1 | 2 - 3 | 78 | |||
| GenBank mRNA | BC075494.1 | - | 2 - 3 | 77 | ||
| BC161333.1 | - | 2 - 3 | 206 | 78 | ||
| CU075356.1 | - | 2 - 3 | 78 | |||
More Information
Set Membership: |
|

ripply3
Genomic Map