Assay Details
| Gene Symbol: ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127017.1 | NP_001120489.1 | 2 - 3 | 394 | 86 | |
| XM_012970101.2 | XP_012825555.1 | 2 - 3 | 330 | 86 | ||
| GenBank mRNA | BC161365.1 | - | 2 - 3 | 394 | 86 | |
More Information
Set Membership: |
|

ubfd1
Genomic Map