Assay Details
| Gene Symbol: sntb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127021.1 | NP_001120493.1 | 4 - 5 | 1092 | 67 | |
| GenBank mRNA | BC161370.1 | - | 4 - 5 | 1092 | 67 | |
More Information
Set Membership: |
|

sntb2
Genomic Map