Assay Details
| Gene Symbol: zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127030.1 | NP_001120502.1 | 6 - 7 | 1568 | 68 | |
| XM_012968033.2 | XP_012823487.1 | 12 - 13 | 2490 | 68 | ||
| XM_012968034.2 | XP_012823488.1 | 12 - 13 | 3068 | 68 | ||
| XM_012968035.2 | XP_012823489.1 | 12 - 13 | 3065 | 68 | ||
| XM_012968036.2 | XP_012823490.1 | 12 - 13 | 2918 | 68 | ||
| GenBank mRNA | BC161391.1 | - | 6 - 7 | 1568 | 68 | |
| BC168044.1 | - | 12 - 13 | 2895 | 68 | ||
More Information
Set Membership: |
|

zc3h4
Genomic Map