Assay Details
| Gene Symbol: usf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127125.1 | NP_001120597.1 | 3 - 4 | 295 | 73 | |
| XM_012956505.2 | XP_012811959.1 | 3 - 4 | 354 | 73 | ||
| GenBank mRNA | BC161632.1 | - | 3 - 4 | 295 | 73 | |
More Information
Set Membership: |
|

usf3
Genomic Map