Assay Details
| Gene Symbol: arc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127127.1 | NP_001120599.1 | 4 - 5 | 552 | 89 | |
| GenBank mRNA | AB474592.1 | - | 3 - 4 | 458 | 89 | |
| BC161634.1 | - | 4 - 5 | 552 | 89 | ||
| CT030302.1 | - | 4 - 5 | 505 | 89 | ||
More Information
Set Membership: |
|

arc3
Genomic Map