Assay Details
| Gene Symbol: wdr83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126516.1 | NP_001119988.1 | 780 | 69 | ||
| GenBank mRNA | BC158245.1 | - | 780 | 69 | ||
| CR942720.2 | - | 3 - 4 | 268 | 69 | ||
More Information
Set Membership: |
|

wdr83os
Genomic Map