Assay Details
| Gene Symbol: sdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126528.1 | NP_001120000.1 | 149 | 66 | ||
| GenBank mRNA | BC158270.1 | - | 149 | 66 | ||
| CR942628.2 | - | 82 | 66 | |||
More Information
Set Membership: |
|

sdhb
Genomic Map