Assay Details
| Gene Symbol: wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126537.1 | NP_001120009.1 | 1 - 2 | 146 | 57 | |
| GenBank mRNA | BC158306.1 | - | 1 - 2 | 146 | 57 | |
More Information
Set Membership: |
|

wdr77
Genomic Map