Assay Details
| Gene Symbol: gid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126573.1 | NP_001120045.1 | 5 - 6 | 730 | 69 | |
| GenBank mRNA | BC158383.1 | - | 5 - 6 | 730 | 69 | |
More Information
Set Membership: |
|

gid4
Genomic Map