Assay Details
| Gene Symbol: dapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126642.1 | NP_001120114.1 | 4 - 5 | 531 | 86 | |
| XM_012966053.2 | XP_012821507.1 | 4 - 5 | 612 | 86 | ||
| XM_012966054.2 | XP_012821508.1 | 4 - 5 | 608 | 86 | ||
| GenBank mRNA | BC158982.1 | - | 4 - 5 | 531 | 86 | |
More Information
Set Membership: |
|

dapk2
Genomic Map