Assay Details
| Gene Symbol: uck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126769.1 | NP_001120241.1 | 4 - 5 | 641 | 59 | |
| GenBank mRNA | BC160433.1 | - | 4 - 5 | 641 | 59 | |
| BC161596.1 | - | 4 - 5 | 591 | 59 | ||
| CT025214.2 | - | 4 - 5 | 524 | 59 | ||
More Information
Set Membership: |
|

uck2
Genomic Map