Assay Details
| Gene Symbol: plppr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126788.1 | NP_001120260.1 | 6 - 7 | 734 | 90 | |
| XM_018090851.1 | XP_017946340.1 | 6 - 7 | 757 | 90 | ||
| GenBank mRNA | BC160501.1 | - | 6 - 7 | 734 | 90 | |
More Information
Set Membership: |
|

plppr3
Genomic Map