Assay Details
| Gene Symbol: trim62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126791.1 | NP_001120263.1 | 527 | 60 | ||
| GenBank mRNA | BC160508.1 | - | 527 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

trim62| Gene Symbol: trim62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126791.1 | NP_001120263.1 | 527 | 60 | ||
| GenBank mRNA | BC160508.1 | - | 527 | 60 | ||
Set Membership: |
|