Assay Details
| Gene Symbol: ntmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126909.1 | NP_001120381.1 | 3 - 4 | 499 | 101 | |
| XM_012968019.2 | XP_012823473.1 | 3 - 4 | 635 | 101 | ||
| XM_012968020.2 | XP_012823474.1 | 3 - 4 | 902 | 101 | ||
| GenBank mRNA | BC161081.1 | - | 3 - 4 | 499 | 101 | |
More Information
Set Membership: |
|

ntmt1
Genomic Map