Assay Details
| Gene Symbol: tgm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126927.1 | NP_001120399.1 | 4 - 5 | 707 | 67 | |
| GenBank mRNA | BC161110.1 | - | 4 - 5 | 707 | 67 | |
More Information
Set Membership: |
|

tgm6
Genomic Map