Assay Details
| Gene Symbol: relch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127054.1 | NP_001120526.1 | 13 - 14 | 1994 | 62 | |
| XM_012964309.2 | XP_012819763.1 | 14 - 15 | 4936 | 62 | ||
| XM_012964310.2 | XP_012819764.1 | 14 - 15 | 4936 | 62 | ||
| XM_012964311.2 | XP_012819765.1 | 14 - 15 | 4936 | 62 | ||
| XM_018094548.1 | XP_017950037.1 | 14 - 15 | 4936 | 62 | ||
| XM_018094549.1 | XP_017950038.1 | 14 - 15 | 4936 | 62 | ||
| GenBank mRNA | BC124571.1 | - | 13 - 14 | 1994 | 62 | |
More Information
Set Membership: |
|

relch
Genomic Map