Assay Details
| Gene Symbol: ptger4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127082.1 | NP_001120554.1 | 1 - 2 | 1343 | 71 | |
| GenBank mRNA | BC161489.1 | - | 1 - 2 | 1343 | 71 | |
More Information
Set Membership: |
|

ptger4
Genomic Map