Assay Details
| Gene Symbol: tigar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127097.1 | NP_001120569.1 | 4 - 5 | 319 | 64 | |
| GenBank mRNA | BC161536.1 | - | 4 - 5 | 319 | 64 | |
| CU075676.1 | - | 4 - 5 | 302 | 64 | ||
More Information
Set Membership: |
|

tigar
Genomic Map