Assay Details
| Gene Symbol: bbox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127102.1 | NP_001120574.1 | 5 - 6 | 909 | 64 | |
| GenBank mRNA | BC161551.1 | - | 5 - 6 | 909 | 64 | |
| CU075844.1 | - | 5 - 6 | 902 | 64 | ||
More Information
Set Membership: |
|

bbox1
Genomic Map