Assay Details
| Gene Symbol: tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127106.1 | NP_001120578.1 | 4 - 5 | 615 | 90 | |
| GenBank mRNA | BC161560.1 | - | 4 - 5 | 615 | 90 | |
More Information
Set Membership: |
|

tcp11l2
Genomic Map