Assay Details
| Gene Symbol: lrrc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127162.1 | NP_001120634.1 | 3 - 4 | 1181 | 79 | |
| GenBank mRNA | BC161789.1 | - | 3 - 4 | 1181 | 79 | |
More Information
Set Membership: |
|

lrrc47
Genomic Map