Assay Details
| Gene Symbol: arl6ip5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127430.1 | NP_001120902.1 | 2 - 3 | 396 | 66 | |
| GenBank mRNA | BC166257.1 | - | 2 - 3 | 396 | 66 | |
More Information
Set Membership: |
|

arl6ip5
Genomic Map