Assay Details
| Gene Symbol: slc9a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127431.1 | NP_001120903.1 | 11 - 12 | 1527 | 65 | |
| XM_012956513.2 | XP_012811967.1 | 11 - 12 | 1647 | 65 | ||
| GenBank mRNA | BC166339.1 | - | 11 - 12 | 1527 | 65 | |
More Information
Set Membership: |
|

slc9a7
Genomic Map