Assay Details
| Gene Symbol: arhgap11a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127991.1 | NP_001121463.1 | 3 - 4 | 625 | 84 | |
| XM_012968063.2 | XP_012823517.1 | 3 - 4 | 829 | 84 | ||
| XM_012968064.2 | XP_012823518.1 | 3 - 4 | 179 | 84 | ||
| GenBank mRNA | BC158291.1 | - | 3 - 4 | 488 | 84 | |
| BC166229.1 | - | 3 - 4 | 625 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012968064.2 transcripts.
Set Membership: |
|

arhgap11a.1
Genomic Map