Assay Details
| Gene Symbol: arhgap11a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127991.1 | NP_001121463.1 | 5 - 6 | 1054 | 94 | |
| XM_012968063.2 | XP_012823517.1 | 5 - 6 | 1258 | 94 | ||
| XM_012968064.2 | XP_012823518.1 | 5 - 6 | 608 | 94 | ||
| GenBank mRNA | BC158291.1 | - | 5 - 6 | 917 | 94 | |
| BC166229.1 | - | 5 - 6 | 1054 | 94 | ||
More Information
Set Membership: |
|

arhgap11a.1
Genomic Map