Assay Details
| Gene Symbol: arhgap11a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127991.1 | NP_001121463.1 | 8 - 9 | 1515 | 80 | |
| XM_012968063.2 | XP_012823517.1 | 9 - 10 | 1794 | 80 | ||
| XM_012968064.2 | XP_012823518.1 | 9 - 10 | 1144 | 80 | ||
| GenBank mRNA | BC166229.1 | - | 8 - 9 | 1515 | 80 | |
More Information
Set Membership: |
|

arhgap11a.1
Genomic Map