Assay Details
| Gene Symbol: upf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128003.1 | NP_001121475.1 | 20 - 21 | 3811 | 65 | |
| XM_012958639.2 | XP_012814093.1 | 20 - 21 | 4051 | 65 | ||
| XM_012958640.2 | XP_012814094.1 | 20 - 21 | 4063 | 65 | ||
| XM_012958641.2 | XP_012814095.1 | 20 - 21 | 4055 | 65 | ||
| GenBank mRNA | BC166247.1 | - | 20 - 21 | 3811 | 65 | |
| BC168584.1 | - | 19 - 20 | 3564 | 65 | ||
| CR855447.2 | - | 3 - 4 | 397 | 65 | ||
More Information
Set Membership: |
|

upf2
Genomic Map