Assay Details
| Gene Symbol: acsbg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128023.1 | NP_001121495.1 | 3 - 4 | 432 | 74 | |
| GenBank mRNA | BC166305.1 | - | 3 - 4 | 432 | 74 | |
More Information
Set Membership: |
|

acsbg1
Genomic Map