Assay Details
| Gene Symbol: rad54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128037.1 | NP_001121509.1 | 7 - 8 | 1188 | 67 | |
| XM_012964352.2 | XP_012819806.1 | 6 - 7 | 1032 | 67 | ||
| GenBank mRNA | BC166345.1 | - | 7 - 8 | 1188 | 67 | |
More Information
Set Membership: |
|

rad54b
Genomic Map