Assay Details
| Gene Symbol: rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128659.1 | NP_001122131.1 | 1 - 2 | 410 | 74 | |
| GenBank mRNA | BC167318.1 | - | 1 - 2 | 410 | 74 | |
| CU075657.1 | - | 1 - 2 | 421 | 74 | ||
More Information
Set Membership: |
|

rhov
Genomic Map