Assay Details
| Gene Symbol: pgm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130233.1 | NP_001123705.1 | 1 - 2 | 298 | 95 | |
| GenBank mRNA | BC167308.1 | - | 1 - 2 | 298 | 95 | |
More Information
Set Membership: |
|

pgm5
Genomic Map