Assay Details
| Gene Symbol: lefty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130253.1 | NP_001123725.1 | 1 - 2 | 323 | 91 | |
| GenBank mRNA | BC167366.1 | - | 1 - 2 | 323 | 91 | |
| CU025005.1 | - | 1 - 2 | 311 | 91 | ||
More Information
Set Membership: |
|

lefty
Genomic Map