Assay Details
| Gene Symbol: larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130268.1 | NP_001123740.1 | 16 - 17 | 2140 | 61 | |
| XM_012964365.2 | XP_012819819.1 | 14 - 15 | 5451 | 61 | ||
| XM_012964367.2 | XP_012819821.1 | 16 - 17 | 2254 | 61 | ||
| GenBank mRNA | BC167405.1 | - | 16 - 17 | 2140 | 61 | |
| CR848356.2 | - | 2 - 3 | 197 | 61 | ||
More Information
Set Membership: |
|

larp4b
Genomic Map