Assay Details
| Gene Symbol: zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130363.1 | NP_001123835.1 | 3 - 4 | 443 | 73 | |
| XM_012960673.2 | XP_012816127.1 | 3 - 4 | 393 | 73 | ||
| XM_018092942.1 | XP_017948431.1 | 3 - 4 | 435 | 73 | ||
| GenBank mRNA | BC167671.1 | - | 3 - 4 | 443 | 73 | |
More Information
Set Membership: |
|

zdhhc24
Genomic Map