Assay Details
| Gene Symbol: runx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135116.2 | NP_001128588.1 | 3 - 4 | 478 | 68 | |
| XM_012970171.2 | XP_012825625.1 | 3 - 4 | 455 | 68 | ||
| GenBank mRNA | AB236909.1 | - | 3 - 4 | 563 | 68 | |
More Information
Set Membership: |
|

runx2
Genomic Map