Assay Details
| Gene Symbol: aarsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142040.1 | NP_001135512.1 | 4 - 5 | 463 | 64 | |
| XM_012971566.2 | XP_012827020.1 | 3 - 4 | 411 | 64 | ||
| XM_012971567.2 | XP_012827021.1 | 4 - 5 | 515 | 64 | ||
| GenBank mRNA | BC167931.1 | - | 4 - 5 | 463 | 64 | |
More Information
Set Membership: |
|

aarsd1
Genomic Map