Assay Details
| Gene Symbol: rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142100.2 | NP_001135572.1 | 2 - 3 | 290 | 64 | |
| XM_012954789.2 | XP_012810243.1 | 2 - 3 | 718 | 64 | ||
| XM_012954790.2 | XP_012810244.1 | 2 - 3 | 718 | 64 | ||
| GenBank mRNA | BC168076.1 | - | 2 - 3 | 292 | 64 | |
More Information
Set Membership: |
|

rnf8
Genomic Map