Assay Details
| Gene Symbol: mcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142123.1 | NP_001135595.1 | 6 - 7 | 895 | 73 | |
| XM_012957487.2 | XP_012812941.1 | 8 - 9 | 1477 | 73 | ||
| XM_012957491.2 | XP_012812945.1 | 8 - 9 | 1477 | 73 | ||
| XM_012957499.2 | XP_012812953.1 | 6 - 7 | 1306 | 73 | ||
| XM_012957501.2 | XP_012812955.1 | 6 - 7 | 936 | 73 | ||
| GenBank mRNA | BC168123.1 | - | 6 - 7 | 895 | 73 | |
More Information
Set Membership: |
|

mcc
Genomic Map