Assay Details
| Gene Symbol: chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142158.1 | NP_001135630.1 | 1110 | 63 | ||
| GenBank mRNA | BC168482.1 | - | 1110 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

chpf2| Gene Symbol: chpf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142158.1 | NP_001135630.1 | 1110 | 63 | ||
| GenBank mRNA | BC168482.1 | - | 1110 | 63 | ||
Set Membership: |
|